IG® sgRNA Synthesis Kit for Cas12a Nuclease, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
3303 3306
104090-508PK 275.77 USD
104090-508 104090-510
IG® sgRNA Synthesis Kit for Cas12a Nuclease, Intact Genomics
Enzymes
It is your resource for in vitro gene editing using Cas12a enzyme. We understand your need to generate sufficient quantities of sgRNA for in vitro gene editing projects. The IG® sgRNA synthesis kits are simple to use, and our kits are scalable to help you reach these goals. The IG® sgRNA synthesis kit for Cas12a Nuclease can be purchased with or without the sgRNA purification kit.
  • IG® sgRNA Synthesis Kit for Cas12a Nuclease includes everything but your targeting oligo (all buffers, enzymes, scaffolds)
  • User simply needs to provide the oligo for your target—Must be 32-38 base pairs and generated using the template design in this manual
  • Rapid Workflow (Less than 1 hour)
  • Scalable Yield (just 1 reaction for all the sgRNA you need)
IG® sgRNA Synthesis Kits are a perfect choice for a variety of sgRNA synthesis needs. Below is a sampling of some of the key benefits. With high and scalable yield, the IG® sgRNA Synthesis Kit for Cas12a nuclease is the perfect choice for a variety of sgRNA synthesis needs.
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.