BL21(DE3) Chemically Competent Cells
Supplier: INTACT GENOMICS, INC MS
1051-48
104090-444PK 444.23 USD
104090-444
BL21(DE3) Chemically Competent Cells
Competent Cells
For strong inducible protein expression under the bacteriophage T7 promoter.
  • Widely used host background
  • T7 Expression Strain
  • Deficient in both lon (1) and ompT proteases
  • Resistant to phage T1 (fhuA2)
  • B Strain
Intact Genomics chemically competent BL21(DE3) E. coli cells are suitable for transformation and routine protein expression.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system