AccuVac® Test Kits, Photometric
Supplier: Hach
2507025 2508025 2515025 2517025 2501025 DRB200-08 2503025 2516025 2518025
104086-362PK 75.94 USD
104086-362 104086-370 104086-372 104086-376 104086-358 104088-462 104088-232 104088-236 104088-238
AccuVac® Test Kits, Photometric
Environmental Test Kits
AV - AccuVac® - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.