Sample Cuvettes
Supplier: Hach
4165802 2427606 4864302
104086-786EA 69.72 USD
104086-786 104088-212 104088-340
Sample Cuvettes
Cuvettes Spectrophotometer Cuvettes
These high range sample cuvettes are designed for use with all spectrophotometers.
Suitable for chlorine applications.