Buffer Solutions for Hardness Testing
Supplier: Hach
42449 42453
104088-328EA 57.58 USD
104088-328 104088-330
Buffer Solutions for Hardness Testing
Environmental Test Kits
This buffer is used for hardness and chelant determination by titration.