Stablcal® Turbidity Standards Calibration Kit for 2100Q
Supplier: Hach
2659400 2659405 2659410 2961701 2971200 2971210 2971205
104086-514EA 888.93 USD
104086-514 104086-516 104086-518 104086-726 104086-736 104086-738 104088-316
Stablcal® Turbidity Standards Calibration Kit for 2100Q
Standards Turbidity Standards
Stablcal® calibration standards kit, for 2100 series portable turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilized formazin turbidity standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labour
  • Confidence in results