Powder Pillows, Chromium
Supplier: Hach
1206699 204399 204499 1271099
104085-236PK 129.08 USD
104085-236 104085-712 104085-714 104088-092
Powder Pillows, Chromium
Environmental Test Kits
ChromaVer 3 reagent for hexavalent chromium determination by the Diphenylcarbohydrazide Method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023.