Human c-MET/HGFR Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
IB30301 IB30302
104087-936KT 1163.08 USD
104087-936 104087-938
Human c-MET/HGFR Luciferase Reporter Assay Kit
Assays Enzyme Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's c-MET/hepatocyte growth factor receptor reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against c-MET (HGFR).
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, luciferase detection reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting c-MET/hepatocyte growth factor receptor (c-MET). INDIGO's c-MET reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the c-MET. In addition to c-MET reporter cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, luciferase detection reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against c-MET. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw. Consistent results lot to lot.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.