TNTplus™ Chromium Kit
Supplier: Hach
TNT854
104087-848PK 180.21 USD
104087-848
TNTplus™ Chromium Kit
Environmental Test Kits
This kit is used for the determination of hexavalent and total chromium by the 1,5-Diphenylcarbohydrazide method using TNTplus™ vials.
  • Easy and safe handling
  • No reagent blank necessary
  • Automatic method detection