TNTplus™ Nitrogen Tests
Supplier: Hach
TNT826 TNT827 TNT828
104087-818PK 190.81 USD
104087-818 104087-820 104087-822
TNTplus™ Nitrogen Tests
Environmental Test Kits
These 'Test N Tube' are used for the determination of nitrogen by the persulfate digestion method.
  • Test 'N Tube products provide safe, convenient testing
  • TNT plus vials reduce errors with bar coded vials and RFID
  • TNT plus vials require no reagent blank