IntelliCAL™ MTC101/MTC301 ORP Combination Electrodes
Supplier: Hach
MTC10101 MTC10103 MTC10105 MTC10110 MTC10115 MTC10130 MTC30101 MTC30103
104087-732EA 1018.86 USD
104087-732 104087-734 104087-736 104087-738 104087-740 104087-742 104087-744 104087-746
IntelliCAL™ MTC101/MTC301 ORP Combination Electrodes
Electrodes Redox Electrodes
Digital combination ORP/Redox probes with built-in temperature sensor. Ideal for measuring absolute mV values in wastewater, drinking water and general applications.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Traceability in measurement history: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • User alerted when re-calibration is needed
  • Plug & Play: Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Rugged digital probes can reliably test in the harshest environmental conditions
Available with a non refillable gel electrolyte (MTC101) or refillable 3M KCl electrolyte (MTC301).
The standard versions are available with a 1 or 3 meter cable and are intended for laboratory use. The rugged versions are available with a 5, 10 or 15 meter cable and are intended for field use.