sensION+ 5070 Conductivity Cell, Platinum
Supplier: Hach
LZW5070970002
104087-712EA 1074.6 USD
104087-712
sensION+ 5070 Conductivity Cell, Platinum
Electrodes Conductivity Sensors
This three-pole platinum cell with an integrated Pt1000 temperature sensor provides accurate, stable results over a wide measurement range.
  • Low maintenance requirements
  • Fixed 1 meter cable and telephonic connector
  • Dedicated for use with Hach sensION+ laboratory conductivity meters
  • Ideal for conductivity measurements in general aqueous applications
Delivery information: Supplied with basic user manual.