sensION+ 5056 ORP Electrode
Supplier: Hach
LZW5056970002
104087-702EA 803.08 USD
104087-702
sensION+ 5056 ORP Electrode
Electrodes Redox Electrodes
sensION+ 5056 is a combination platinum ORP electrode with a glass body and non refillable gel-electrolyte reference. It is dedicated for use with Hach sensION+ labpratory pH (mV) meters.
  • Fixed 1 meter cable and BNC connector
  • Low-maintenance design for a variety of applications
  • Gel-electrolyte reference and platinum wire sensor provides accurate, stable results over a wide range of applications
  • Encapsulated cartridge reference system ensures premium stability and long lifespan
  • Innovative reference system utilises a “cartridge” of Ag/AgCl crystals contacting a silver wire inside a glass tube
The 5056 is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.
Delivery information: Supplied with basic user manual.