sensION+ 5048 Multi-Parameter Electrode (pH/ORP/Conductivity)
Supplier: Hach
LZW5048970002
104087-692EA 1583.64 USD
104087-692
sensION+ 5048 Multi-Parameter Electrode (pH/ORP/Conductivity)
Electrodes Multi-Parameter Electrodes
Multisensor for pH, ORP, conductivity and temperature. For use with sensION+ MM150 multi-parameter meter.
  • Accurate
  • Easy to use
  • Rugged and durable
  • Automatic temperature compensation
  • Glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors
Attachable calibration tubes can screw into handle of electrode (only 2 ml of buffer or standard). Storage protector prevents breakage and avoids over pressurisation of the diaphragm. Ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.