sensION+ 5045 Multi-Parameter Electrode (pH/ORP/Temperature)
Supplier: Hach
LZW5045970002
104087-690EA 1246.06 USD
104087-690
sensION+ 5045 Multi-Parameter Electrode (pH/ORP/Temperature)
Electrodes Multi-Parameter Electrodes
sensION+ 5045 is a combination multi-parameter electrode for determination of pH, ORP and temperature. Dedicated for use with Hach sensION+ MM110 portable meters.
  • Ceramic pin reference junction and glass (pH); platinum wire (ORP); and Pt1000 (temperature) sensors
  • Plastic body, non refillable gel reference electrolyte and built-in temperature sensor
  • Low maintenance design for a variety of applications
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
  • Innovative designs house multiple sensing elements in a standard 12 mm electrode body
The electrode is ideal for pH, ORP and temperature measurements in general aqueous applications.