sensION+ 5044 Reference Electrode for Use with Ion Selective Electrodes
Supplier: Hach
LZW5044970002
104087-688EA 668.77 USD
104087-688
sensION+ 5044 Reference Electrode for Use with Ion Selective Electrodes
Electrodes ISE Electrodes
The 5044 with lithium acetate electrolyte is recommended for use with all sensION+ half-cell ion selective electrodes (ISE) in drinking water, wastewater and general water quality applications.
  • Double junction reference electrode with internal gel electrolyte and 0,1 M lithium acetate salt bridge
  • Guarded against contamination
  • Quick measurement stabilization with large ceramic junction
  • Easy to maintain: Lithium acetate electrolyte is refillable
  • Fixed 1 meter cable and banana connector for laboratory use with the Hach sensION+ MM374 Multimeter