Supplier: Hach
LQV1489900002
104087-498EA 2242.8 USD
104087-498
Shakers and Mixers Orbital Shakers
The TOC X5 Shaker is required for the TNT810 and TNT811 TOC TNTplus tests. It purges the Total Inorganic Carbon in the vial, which means you do not have to use or clean any lab glassware.
  • Mains Connection 3: 30 VA
  • Manual Languages: English, French, Spanish, Portugese, Chinese
  • Parameter: TOC (Total Organic Carbon)
  • Warranty: 12 months
The TOC X5 Shaker allows to prepare eight samples in parallel in only five minutes, shortening the time spent on your analysis.
Delivery information: Includes: TOC-X5 Shaker, plug-in power supply unit, non-slip mat.