IntelliCAL™ Luminescent/Optical Dissolved Oxygen (LDO) Electrodes
Supplier: Hach
LDO10101 LDO10103 LDO10105 LDO10115 LDO10130
104087-354EA 2107.8 USD
104087-354 104087-356 104087-358 104087-360 104087-362
IntelliCAL™ Luminescent/Optical Dissolved Oxygen (LDO) Electrodes
Electrodes Dissolved Oxygen Electrodes
IntelliCAL™ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
  • Rugged digital probes can reliably test in the harshest environmental conditions: Stainless steel body, polymer sensor shroud, and reinforced, steel-sheathed cables protect the electrode for measurement anywhere in the field
  • No membranes to clean or replace, no electrolyte solution to replenish, and reduce calibration frequency
  • Ultimate traceability: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Alert the user when re-calibration is needed
  • Electrodes can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Ideal for a multi-user environment with multiple HQd series meters
The standard version is intended for laboratory use, the rugged version is intended for field use.