pH/mV/°C Meter, Bench, HQd (HQ411D)
Supplier: Hach
HQ411D
104087-304EA 2385.76 USD
104087-304
pH/mV/°C Meter, Bench, HQd (HQ411D)
Electrochemistry Meters pH and Multi-Parameter Meters
This advanced laboratory meter is ideal for testing critical water quality parameters. The meter connects with smart probes (IntelliCAL™) that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The instrument is designed to withstand years of continuous lab use.
  • Single input channel for pH, ORP, mV, and temperature: For use with any IntelliCAL™ smart pH or ORP electrode
  • Simple user interface that does not require manuals or training to operate
  • Guided calibration and check standard routines reduce calibration errors
  • Internal USB ports simplify data transfer, peripherals, and software updates and connection to PCs printer and keyboards
  • Fully GLP compliant communication and documentation
  • Ultra-bright screen and large font size for easy to read results
Ordering information: Supplied with four batteries, universal power adapter and with or without electrode holder. IntelliCAL™ electrodes need to be ordered separately.