Radiometer Analytical Conductivity Cells
Supplier: Hach
B15B001 E61M008 E61M009 E61M010 E61M011 E61M012 E61M013 E61M016 E61M022 B60E100 E61M015
104087-164EA 1844.31 USD
104087-164 104087-266 104087-268 104087-270 104087-272 104087-274 104087-276 104087-278 104087-280 104088-444 104088-474
Radiometer Analytical Conductivity Cells
Electrodes Conductivity Sensors
Range of conductivity cells for a wide variety of applications.
  • Two-pole cells: traditional design based on two plates of platinum; ideal for routine measurement of conductivity and for use with a sample changer due to the easy rinsing
  • Three-pole cells: three platinum rings which facilitate optimal shielding during measurement
  • Four-pole cells: four platinum rings ensure accurate results over several decades of conductivity with a single cell using just one calibration
  • four-pole cells are particularly recommended when performing high conductivity measurements