Salt Bridges
Supplier: Hach
B40A520
104087-176EA 195.23 USD
104087-176
Salt Bridges
Tubes Test Tubes
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.