Radiometer Analytical Reference Electrodes
Supplier: Hach
B20B300 E21M001 E21M002 E21M009
104087-166EA 621.69 USD
104087-166 104088-466 104088-468 104088-470
Radiometer Analytical Reference Electrodes
Electrodes ISE Electrodes
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
  • Liquid junction: porous pin
  • Red-Rod technology provides a reproducible, fast and accurate response
  • Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.