Chemkey® Reagents
Supplier: Hach
8636200
8636300
8636400
8636500
8636600
8791900
9429000
9429100
9429200
9429300
8636000
8636100
9425200
8499300
8499400
9429400
9429500
9429600
104086-978BX
46.17
USD
104086-978
104086-980
104086-982
104086-984
104086-986
104086-994
104087-060
104087-062
104087-064
104087-066
104086-974
104086-976
104087-056
104086-926
104086-928
104087-068
104087-070
104087-072
Chemkey® Reagents
Environmental Test Kits
Chemkey® reagents are a new technology that enable the SL1000 Portable Parallel Analyzer colorimeter to perform the same tests with less than half the manual steps.
- All chemicals and processes are entirely contained inside the Chemkey®
- There are no powder pillows or glass vials to handle
- Get highly accurate results with less opportunity for errors in a fraction of the time
There is no zeroing, no mixing, no shaking, no chemicals or vials to handle. All you have to do is insert the chemkey and let the PPA-SL1000 read you back the results.