Chemkey® Reagents
Supplier: Hach
8636200 8636300 8636400 8636500 8636600 8791900 9429000 9429100 9429200 9429300 8636000 8636100 9425200 8499300 8499400 9429400 9429500 9429600
104086-978BX 46.17 USD
104086-978 104086-980 104086-982 104086-984 104086-986 104086-994 104087-060 104087-062 104087-064 104087-066 104086-974 104086-976 104087-056 104086-926 104086-928 104087-068 104087-070 104087-072
Chemkey® Reagents
Environmental Test Kits
Chemkey® reagents are a new technology that enable the SL1000 Portable Parallel Analyzer colorimeter to perform the same tests with less than half the manual steps.
  • All chemicals and processes are entirely contained inside the Chemkey®
  • There are no powder pillows or glass vials to handle
  • Get highly accurate results with less opportunity for errors in a fraction of the time
There is no zeroing, no mixing, no shaking, no chemicals or vials to handle. All you have to do is insert the chemkey and let the PPA-SL1000 read you back the results.