Test Kits, Drop Count Tests
Supplier: Hach
143601 145700 145701 2291700 2308100 2687200 145201 145300 145400 208601 225401 2437500 2437600 2437700 2437800 2440000 144001 146900 2408549 222702 42632 2444300 148002 2314500 2183100
104085-424EA 135.81 USD
104085-424 104085-508 104085-510 104086-084 104086-096 104086-576 104085-482 104085-484 104085-488 104085-752 104086-010 104086-262 104086-264 104086-266 104086-268 104086-272 104085-456 104085-528 104086-194 104085-932 104086-794 104086-276 104088-116 104088-204 104088-170
Test Kits, Drop Count Tests
Environmental Test Kits
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.