Starch Indicator Solution, MDB
Supplier: Hach
34932
104086-774EA 33.88 USD
104086-774
Starch Indicator Solution, MDB
Environmental Test Kits
Oxidation-reduction indicator for the analysis of BOD, total chlorine, chromium, cyanide, dissolved oxygen and sulfite.
  • Marked dropping bottle (MDB)