Potassium iodide solution 30 g/L

Supplier: Hach

Synonyms: Dipotassium diiodide, Kali iodide

34332
97026-676EA 42.21 USD
97026-676 104086-772
Potassium iodide solution 30 g/L
Potassium iodide

Reagent for use in iodometric titrations.


Method: 8210

Formula: Kl
MW: 166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR