Sodium thiosulfate 0.1 mol/L (0.1 N) in aqueous solution

Supplier: Hach

32332
104086-758EA 41.79 USD
104086-758
Sodium thiosulfate 0.1 mol/L (0.1 N) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR