Filling Solution for ISE Electrodes, Sodium
Supplier: Hach
2965126
104086-730EA 62.71 USD
104086-730
Filling Solution for ISE Electrodes, Sodium
Electrolytes Electrode Filling Solutions
Internal filling solution for use with ISENA381 IntelliCAL™ Ultra sodium ion selective electrodes.