BART Microorganism Tests, Test for Acid Producing Bacteria
Supplier: Hach
2831409
104086-700PK 377.68 USD
104086-700
BART Microorganism Tests, Test for Acid Producing Bacteria
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment