Potassium hydroxide standard solution 8.00 N

Supplier: Hach
Danger

28232H
97026-728EA 35 USD
97026-728 104086-692
Potassium hydroxide standard solution 8.00 N
Potassium hydroxide

For pH adjustment in calcium determination by EDTA titration


  • MDB botle


Hach methods 8222 (buret) and 8204 (Digital Titrator).

Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR