Contact Plates
Supplier: Hach
2811615
104086-682PK 268.82 USD
104086-682
Contact Plates
Media for Microbiology Contact Plates for Microbiology
m-Endo prepared agar plates for detection of total coliform bacteria.