Stablcal® Turbidity Standard, ULR Verification Kit
Supplier: Hach
2714600
104086-596EA 391.38 USD
104086-596
Stablcal® Turbidity Standard, ULR Verification Kit
Standards Turbidity Standards
Stablcal® verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 ml each), sililcone oil, oiling cloth and instructions is used for verification purpose only.
  • Precise, pre-mixed formazin solutions
  • Enhanced stability saves time and labor
  • Confidence in results