Phenol red solution, Spectrophotometric Grade pH-indicator

Supplier: Hach

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

2657512
97026-490EA 37.9 USD
97026-490 104086-510
Phenol red solution, Spectrophotometric Grade pH-indicator
Phenol red

Phenol Red Indicator Solution


Standardized for colorimetric pH determination. Color range: pH 6.0-8.5. Bottle/50 mL spec. grade.

Formula: C₁₉H₁₄O₅S
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8

Order Now

Specification Test Results

Method Name Colorimetric Phenol Red
Number of tests 50 mL
Parameter pH
Platform Solution
Quantity 50
Range 6.5 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR