Total Nitrogen Reagent Sets, HR, TNT
Supplier: Hach
2604545 2604945 2672245 TNT822 2606945
104086-468PK 332.08 USD
104086-468 104086-470 104086-566 104088-658 104088-252
Total Nitrogen Reagent Sets, HR, TNT
Environmental Test Kits
For determination of Total Nitrogen by the Persulfate Digestion Test 'N Tube method:
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
  • Perform digestions and photometric measurements in the same vial
Ordering information: 25 to 50 tests (dependent upon the number of reagent blank tests performed).
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.