Visual colour kits, glutaraldehyde
Supplier: Hach
2587200
104086-434EA 1001.56 USD
104086-434
Visual colour kits, glutaraldehyde
Environmental Test Kits
Glutaraldehyde test kits are designed to monitor disinfection processes.
  • Simple, economical colour disc method
  • Accurate and quick results
  • Easy-to-carry test kit
Delivery information: Kits includes three reagents, colour discs, colour comparator box, bottles, sprinkler caps, syringes, cylinders, marker, ampoule breaker, ultra pure water, viewing tubes, instruction sheet, and carrying case.