MEL/m-ColiBlue24® Field Filtration Kit
Supplier: Hach
2586400
104086-432EA 4387.85 USD
104086-432
MEL/m-ColiBlue24® Field Filtration Kit
Microbiology Test Systems Microorganism Tests
The coliform and E.coli field filtration kit features m-ColiBlue24® broth and Millipore's portable filter assembly, push fit funnels and Millipore membranes to simplify membrane filtration testing in the field and also m-ColiBlue24® broth simultaneously detects total coliforms and E.coli in 24 hours.
  • Efficient field operations
  • Includes consumables and media
  • EPA approved field method
Delivery information: The carrying case contains 40 m-ColiBLue24® broth ampules, 150 push fit funnels and S-Pak filters (Millipore), 1 microfil stand alone support (Millipore), 1 syringe/tubing vacuum device, 50 petri dishes (50 mm, sterile) with nutrient pad, 50 sterile, individually wrapped 10 ml pipets (poly), 50 sterile sample bags with dechlorinating agent, alcohol burner and 100 ml of isopropyl alcohol, illuminated magnifier (5X), pipette filler, forceps, ampule breaker and procedure documentation.