Bromothymol blue solution indicator

Supplier: Hach

25532
89230-862EA 43.6 USD
89230-862 104086-398
Bromothymol blue solution indicator
Bromothymol blue

Bromthymol Blue Indicator Solution. Standardized for colorimetric pH determination. pH range 5.5-8.5.


Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.

Formula: C₂₇H₂₈Br₂O₅S
MDL Number: MFCD00005872
CAS Number: 76-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR