Comparator Kits for Water Testing
Supplier: Hach
1416100 1419301 145401 2064400 146700 2518050
104085-358EA 275.64 USD
104085-358 104085-380 104085-490 104085-732 104085-524 104086-378
Comparator Kits for Water Testing
Environmental Test Kits
Qualitative results using traditional color comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.