SpecCheck Secondary Gel Standards Sets, DPD Chlorine, LR
Supplier: Hach
2635300 2507500 2708000 2712500 2893300
97026-500EA 484.46 USD
97026-500 MSPP-2635300 104086-368 104086-588 104086-592 104088-312
SpecCheck Secondary Gel Standards Sets, DPD Chlorine, LR
Standards Environmental Analysis Standards
DPD chlorine, low range, 0 to 2 mg/L as Cl₂.
  • Fast and convenient
  • Stable
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Ordering information: Set of four jars.