Sodium hydroxide 5 mol/L (5 N) in aqueous solution

Supplier: Hach
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

245026
104086-310EA 40.66 USD
104086-310
Sodium hydroxide 5 mol/L (5 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR