Sulfuric acid 2.625 mol/l (5.25 N) in aqueous solution

Supplier: Hach
Danger

244903
104086-300EA 168.92 USD
104086-300
Sulfuric acid 2.625 mol/l (5.25 N) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR