Visual Color Kits, Alkalinity, AL-AP
Supplier: Hach
2444301
104086-278EA 145.73 USD
104086-278
Visual Color Kits, Alkalinity, AL-AP
Environmental Test Kits
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.
  • Kit utilises drop count titration method
  • Simple, economical titration method
Delivery information: Rugged kit contains all required reagents, solutions, and glassware.