Aluminium Reagent Set for Al Determination by Photometric ECR Method
Supplier: Hach
2443400
104086-274EA 439.79 USD
104086-274
Aluminium Reagent Set for Al Determination by Photometric ECR Method
Environmental Test Kits
Kit used for the determination of aluminum by Eriochrome Cyanine R (ECR) method and has sample size of 50 ml.
Delivery information: Set contains ECR reagent powder pillows, hexamethylenetetramine buffer reagent powder pillows and ECR masking reagent solution for approximately 100 tests.