BART Microorganism Tests, Test for Sulfate-Reducing Bacteria
Supplier: Hach
2432427
104086-246PK 1088.4 USD
104086-246
BART Microorganism Tests, Test for Sulfate-Reducing Bacteria
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Sulfate reducing bacteria (SRB) are one of the most important agents in biofouling and can cause corrosion, clogging, fouling of the water and increased hygiene risks.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment