COD Digestion Vials
Supplier: Hach
2415915 2415925 2623425 2125915 2125925 2125815 2125825
104086-210PK 502.62 USD
104086-210 104086-212 104086-496 104085-854 104085-856 104088-164 104088-166
COD Digestion Vials
Environmental Test Kits
Prepared COD reagents eliminate the need for measurement of corrosive and toxic chemicals. Just add sample to the COD vial, digest and measure. Photometric measurements provide accurate results without the equipment and effort required for titrations.
  • Low Range (LR) and High Range (HR) vials are approved by the USEPA for reporting of wastewater COD concentrations
  • Reagent contains mercury - please dispose of according to local regulations
  • COD2 UHR is a mercury-free digestion vial