Sodium thiosulfate solution_SC 0.0250 N Stabilised

Supplier: Hach

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

2409353
89231-070EA 74.26 USD
89231-070 104086-206
Sodium thiosulfate solution_SC 0.0250 N Stabilised
Sodium thiosulphate

For dissolved oxygen determination by Winkler titration


Stabilized sodium thiosulfate formulation.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR