Sodium thiosulfate 0.0113 mol/L (0.0113 N) in aqueous solution

Supplier: Hach

2409032
104086-198EA 53.3 USD
104086-198
Sodium thiosulfate 0.0113 mol/L (0.0113 N) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR