AccuVac® Ampoule Snapper
Supplier: Hach
2405200
104086-192EA 54.73 USD
104086-192
AccuVac® Ampoule Snapper
Ampule Openers
This snapper is used for opening ampoules immersed in samples.
  • Avoids sample contact
  • Used with colorimeters and spectrophotometers