Dissolved Oxygen, Reagent Set
Supplier: Hach
2351400
104086-136EA 257.67 USD
104086-136
Dissolved Oxygen, Reagent Set
Environmental Test Kits
For determination of dissolved oxygen by the Winkler method (azide modification) and buret titration.